siRNA Details
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107067.2 | NP_001100537.2 | 3 | 314 | ||
NM_001270834.1 | NP_001257763.1 | 3 | 242 | |||
GenBank mRNA | FQ222541.1 | - | 3 | 242 | ||
FQ230904.1 | - | 3 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
