siRNA Details
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031652.1 | NP_001026822.1 | 9 | 1000 | ||
GenBank mRNA | AJ634457.1 | - | 9 | 970 | ||
BC099812.1 | - | 9 | 1000 | 7929942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
