siRNA Details
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247938.2 | XP_006248000.1 | 8 | 1895 | ||
XM_008768486.1 | XP_008766708.1 | 9 | 4193 | |||
XM_221188.8 | XP_221188.5 | 8 | 1892 |

More Information
Other Species Targeted

Gene Ontology Categories:
