siRNA Details
Gene Symbol: Slc38a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727846.4 | XP_002727892.2 | 8 | 1049 | ||
XM_006247943.2 | XP_006248005.1 | 8 | 1049 | |||
XM_006247944.2 | XP_006248006.1 | 8 | 1050 | |||
XM_006247945.2 | XP_006248007.1 | 8 | 1049 | |||
XM_006247946.2 | XP_006248008.1 | 8 | 1049 | |||
XM_221195.8 | XP_221195.5 | 8 | 1049 |
