siRNA Details
Gene Symbol: Mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029900.1 | NP_001025071.1 | 4 | 683 | ||
GenBank mRNA | FQ228179.1 | - | 4 | 497 | ||
FQ233456.1 | - | 4 | 499 | |||
FQ234892.1 | - | 4 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
