siRNA Details
Gene Symbol: Fn3krp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107077.2 | NP_001100547.2 | 6 | 746 | ||
GenBank mRNA | BC166839.1 | - | 711 | 9027722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
