siRNA Details
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198760.1 | NP_942055.1 | 5 | 757 | ||
XM_006247613.2 | XP_006247675.1 | 6 | 3435 | |||
XM_006247614.2 | XP_006247676.1 | 6 | 1456 | |||
XM_006247617.2 | XP_006247679.1 | 7 | 1140 | |||
XM_006247618.2 | XP_006247680.1 | 6 | 1022 | |||
XM_006247619.2 | XP_006247681.1 | 5 | 2978 | |||
XM_006247620.2 | XP_006247682.1 | 5 | 2659 | |||
XM_008768375.1 | XP_008766597.1 | 5 | 1268 | |||
XM_008768376.1 | XP_008766598.1 | 6 | 3166 | |||
GenBank mRNA | AY325107.1 | - | 5 | 757 | ||
BC081999.1 | - | 5 | 818 | 7131044 | ||
FQ212616.1 | - | 1 | 92 |
