siRNA Details
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198760.1 | NP_942055.1 | 2 | 271 | ||
XM_006247613.2 | XP_006247675.1 | 3 | 2949 | |||
XM_006247614.2 | XP_006247676.1 | 3 | 970 | |||
XM_006247617.2 | XP_006247679.1 | 4 | 654 | |||
XM_006247618.2 | XP_006247680.1 | 3 | 536 | |||
XM_006247619.2 | XP_006247681.1 | 2 | 2492 | |||
XM_006247620.2 | XP_006247682.1 | 2 | 2173 | |||
XM_008768375.1 | XP_008766597.1 | 2 | 782 | |||
XM_008768376.1 | XP_008766598.1 | 3 | 2680 | |||
GenBank mRNA | AY325107.1 | - | 2 | 271 | ||
BC081999.1 | - | 2 | 332 | 7131044 |
