siRNA Details
Gene Symbol: Dgcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107080.2 | NP_001100550.1 | 2 | 197 | ||
XM_006248609.2 | XP_006248671.1 | 2 | 226 | |||
GenBank mRNA | BC158828.1 | - | 2 | 197 | 7378165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
