siRNA Details
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039021.1 | NP_001034110.1 | 10 | 1294 | ||
XM_006248610.2 | XP_006248672.1 | 10 | 1293 | |||
GenBank mRNA | AY871204.1 | - | 10 | 1294 |

More Information
Other Species Targeted

Gene Ontology Categories:
