siRNA Details
Gene Symbol: Dvl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107081.2 | NP_001100551.1 | 13 | 1303 | ||
XM_006248564.2 | XP_006248626.1 | 13 | 1534 | |||
XM_006248565.2 | XP_006248627.1 | 13 | 1459 | |||
XM_006248566.2 | XP_006248628.1 | 13 | 1459 |
