siRNA Details
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107082.1 | NP_001100552.1 | 9, 10 | 1113 | ||
XM_008768798.1 | XP_008767020.1 | 9, 10 | 1126 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107082.1 | NP_001100552.1 | 9, 10 | 1113 | ||
XM_008768798.1 | XP_008767020.1 | 9, 10 | 1126 |