siRNA Details
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012016.1 | NP_001012016.1 | 6 | 1299 | ||
XM_006248545.2 | XP_006248607.1 | 5 | 1273 | |||
XM_006248546.2 | XP_006248608.1 | 5 | 1421 | |||
GenBank mRNA | BC079273.1 | - | 6 | 1329 | ||
BC089823.1 | - | 6 | 1299 | 7133137 |
