siRNA Details
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107087.1 | NP_001100557.1 | 3 | 575 | ||
XM_008768725.1 | XP_008766947.1 | 3 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
