siRNA Details
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191659.1 | NP_001178588.1 | 6 | 1066 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191659.1 | NP_001178588.1 | 6 | 1066 |