siRNA Details
Gene Symbol: Ildr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127536.3 | NP_001121008.2 | 4 | 477 | ||
XM_006248394.2 | XP_006248456.1 | 4 | 546 | |||
GenBank mRNA | BC166709.1 | - | 4 | 477 | 7929589 |

More Information
Other Species Targeted

Gene Ontology Categories:
