siRNA Details
Gene Symbol: Upk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024253.1 | NP_001019424.1 | 3 | 211 | ||
XM_006248367.2 | XP_006248429.1 | 3 | 418 | |||
XM_006248368.2 | XP_006248430.1 | 3 | 258 | |||
GenBank mRNA | BC093613.1 | - | 3 | 211 | 7309649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
