siRNA Details
Gene Symbol: Hoxd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107094.1 | NP_001100564.1 | 1 | 360 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013554, BC048690, BC013463, AK134330.
Gene Ontology Categories:
