siRNA Details
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107096.1 | NP_001100566.1 | 9, 10 | 1831 | ||
XM_006248250.1 | XP_006248312.1 | 7, 8 | 1506 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107096.1 | NP_001100566.1 | 9, 10 | 1831 | ||
XM_006248250.1 | XP_006248312.1 | 7, 8 | 1506 |