siRNA Details
Gene Symbol: Cpox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037095.1 | NP_001032172.1 | 3 | 836 | ||
GenBank mRNA | BC107664.1 | - | 3 | 836 | 7936479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
