siRNA Details
Gene Symbol: Zfp295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107105.1 | NP_001100575.1 | 4 | 2143 | ||
XM_006248166.2 | XP_006248228.1 | 3 | 2772 | |||
XM_006248167.2 | XP_006248229.1 | 3 | 3207 | |||
XM_006248169.2 | XP_006248231.1 | 3 | 2674 | |||
XM_006248172.2 | XP_006248234.1 | 2 | 2629 | |||
XM_006248174.2 | XP_006248236.1 | 2 | 2531 | |||
XM_006248176.2 | XP_006248238.1 | 4 | 2179 | |||
XM_008768579.1 | XP_008766801.1 | 2 | 7658 |

More Information
Other Species Targeted

Gene Ontology Categories:
