siRNA Details
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107107.1 | NP_001100577.2 | 6 | 802 | ||
XM_006248177.2 | XP_006248239.1 | 6 | 1020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
