siRNA Details
Gene Symbol: Son | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170327.1 | NP_001163798.1 | 3 | 5401 | ||
NM_001170328.1 | NP_001163799.1 | 3 | 5401 | |||
XM_006248064.2 | XP_006248126.1 | 3 | 5419 | |||
XM_006248065.2 | XP_006248127.1 | 3 | 5419 | |||
XM_008768580.1 | XP_008766802.1 | 3 | 5419 | |||
GenBank mRNA | FQ225699.1 | - | 1 | 1197 |

More Information
Other Species Targeted

Gene Ontology Categories:
