siRNA Details
Gene Symbol: Snapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013121.1 | NP_001013139.1 | 5 | 851 | ||
XM_008768956.1 | XP_008767178.1 | 5 | 954 | |||
GenBank mRNA | BC079077.1 | - | 5 | 851 | 7122319 | |
FQ214278.1 | - | 4 | 681 |
