siRNA Details
Gene Symbol: Mmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037217.1 | NP_001032294.1 | 7 | 778 | ||
GenBank mRNA | BC161816.1 | - | 7 | 756 | 7320318 | |
CO404759.1 | - | 4 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
