siRNA Details
Gene Symbol: LOC100363332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769060.1 | XP_008767282.1 | 13 | 2066 |

More Information
Other Species Targeted

Gene Ontology Categories:
