siRNA Details
Gene Symbol: Iqce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248962.2 | XP_006249024.1 | 19 | 1930 | ||
XM_008769035.1 | XP_008767257.1 | 19 | 1930 | |||
XM_008769036.1 | XP_008767258.1 | 18 | 1793 | |||
XM_008769037.1 | XP_008767259.1 | 15 | 1591 | |||
XM_008769038.1 | XP_008767260.1 | 19 | 1928 | |||
XM_221965.7 | XP_221965.5 | 19 | 1930 |
