siRNA Details
Gene Symbol: RGD1566386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107127.2 | NP_001100597.2 | 4 | 1778 | ||
XM_008768973.1 | XP_008767195.1 | 4 | 1782 | |||
GenBank mRNA | BC167756.1 | - | 4 | 1778 | 7099544 |
