siRNA Details
Gene Symbol: Mblac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024996.1 | NP_001020167.1 | 1 | 452 | ||
GenBank mRNA | BC079097.1 | - | 1 | 452 | 7124205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
