siRNA Details
Gene Symbol: Mblac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024996.1 | NP_001020167.1 | 1 | 463 | ||
GenBank mRNA | BC079097.1 | - | 1 | 463 | 7124205 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mblac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024996.1 | NP_001020167.1 | 1 | 463 | ||
GenBank mRNA | BC079097.1 | - | 1 | 463 | 7124205 |