siRNA Details
Gene Symbol: RGD1559588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249038.2 | XP_006249100.1 | 431 | |||
XM_006249039.2 | XP_006249101.1 | 441 | ||||
XM_006249040.2 | XP_006249102.1 | 216 | ||||
XM_221990.6 | XP_221990.4 | 2 | 431 |
Gene Symbol: LOC685048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001062063.5 | XP_001062063.3 | 2 | 431 | ||
XM_006249033.2 | XP_006249095.1 | 2 | 431 | |||
XM_006249034.2 | XP_006249096.1 | 2 | 441 | |||
XM_006249035.2 | XP_006249097.1 | 2 | 240 |
