siRNA Details
Gene Symbol: Gigyf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107133.1 | NP_001100603.1 | 19 | 2424 | ||
XM_006249150.2 | XP_006249212.1 | 16 | 2186 | |||
XM_006249151.1 | XP_006249213.1 | 16 | 2183 | |||
XM_006249152.1 | XP_006249214.1 | 16 | 2183 | |||
XM_006249153.1 | XP_006249215.1 | 17 | 2045 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031408, BC052875, AY176042, AK220289.
Gene Ontology Categories:
