siRNA Details
Gene Symbol: Gigyf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107133.1 | NP_001100603.1 | 24 | 3252 | ||
XM_006249150.2 | XP_006249212.1 | 21 | 3014 | |||
XM_006249151.1 | XP_006249213.1 | 21 | 3011 | |||
XM_006249152.1 | XP_006249214.1 | 21 | 3011 | |||
XM_006249153.1 | XP_006249215.1 | 22 | 2873 |
