siRNA Details
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107137.1 | NP_001100607.1 | 13, 14 | 1800 | ||
GenBank mRNA | BC168924.1 | - | 13, 14 | 1782 | 9029270 |
Gene Symbol: LOC103690026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249229.2 | XP_006249291.2 | 453 |
