siRNA Details
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107137.1 | NP_001100607.1 | 11 | 1473 | ||
GenBank mRNA | BC168924.1 | - | 11 | 1455 | 9029270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
