siRNA Details
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769205.1 | XP_008767427.1 | 6, 7 | 741 | ||
XM_222147.8 | XP_222147.4 | 6, 7 | 596 |

More Information
Other Species Targeted

Gene Ontology Categories:
