siRNA Details
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191665.1 | NP_001178594.1 | 5, 6 | 1182 | ||
XM_006249294.1 | XP_006249356.1 | 5, 6 | 1446 | |||
XM_006249295.1 | XP_006249357.1 | 5, 6 | 1446 | |||
XM_006249296.2 | XP_006249358.1 | 5, 6 | 1446 | |||
XM_006249297.2 | XP_006249359.1 | 5, 6 | 1446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
