siRNA Details
Gene Symbol: Pitpnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107139.1 | NP_001100609.1 | 6 | 1003 | ||
XM_008769227.1 | XP_008767449.1 | 4 | 730 | |||
GenBank mRNA | BC168908.1 | - | 6 | 1088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
