siRNA Details
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249386.2 | XP_006249448.1 | 17 | 2772 | ||
GenBank mRNA | BC168650.1 | - | 17 | 2807 | 7121002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
