siRNA Details
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249386.2 | XP_006249448.1 | 16 | 2639 | ||
GenBank mRNA | BC168650.1 | - | 16 | 2674 | 7121002 |

More Information
Other Species Targeted

Gene Ontology Categories:
