siRNA Details
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191667.1 | NP_001178596.1 | 3 | 689 | ||
XM_008769234.1 | XP_008767456.1 | 3 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
