siRNA Details
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107148.2 | NP_001100618.1 | 3 | 213 | ||
XM_006249542.2 | XP_006249604.1 | 3 | 649 | |||
XM_006249543.2 | XP_006249605.1 | 3 | 646 | |||
XM_006249544.1 | XP_006249606.1 | 4 | 665 | |||
XM_006249545.2 | XP_006249607.1 | 3 | 649 | |||
XM_006249546.2 | XP_006249608.1 | 3 | 649 | |||
XM_008769329.1 | XP_008767551.1 | 4 | 729 | |||
GenBank mRNA | BC168901.1 | - | 3 | 213 | 7385143 |
