siRNA Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122644.1 | NP_001116116.1 | 2 | 666 | ||
XM_008769333.1 | XP_008767555.1 | 2 | 700 | |||
XM_008769334.1 | XP_008767556.1 | 2 | 665 |

More Information
Other Species Targeted

Gene Ontology Categories:
