siRNA Details
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122644.1 | NP_001116116.1 | 2, 3 | 743 | ||
XM_008769333.1 | XP_008767555.1 | 2, 3 | 777 | |||
XM_008769334.1 | XP_008767556.1 | 2, 3 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
