siRNA Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107157.1 | NP_001100627.1 | 12 | 2201 | ||
XM_006249158.1 | XP_006249220.1 | 11 | 2147 | |||
XM_006249159.1 | XP_006249221.1 | 10 | 2009 | |||
XM_008769137.1 | XP_008767359.1 | 11 | 2002 | |||
GenBank mRNA | AY724523.1 | - | 9 | 1844 | ||
BC166738.1 | - | 11 | 2088 | 7937292 |
