siRNA Details
Gene Symbol: Orai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170403.1 | NP_001163874.1 | 3 | 541 | ||
GenBank mRNA | BC158835.1 | - | 3 | 445 | 7384003 | |
EU644750.1 | - | 3 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
