siRNA Details
Gene Symbol: RGD1306072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255328.2 | XP_006255390.1 | 6 | 1287 | ||
XM_006255329.2 | XP_006255391.1 | 7 | 1389 | |||
XM_006255331.2 | XP_006255393.1 | 6 | 1283 | |||
XM_006255335.2 | XP_006255397.1 | 5 | 922 | |||
XM_008772426.1 | XP_008770648.1 | 6 | 1837 | |||
XM_008772427.1 | XP_008770649.1 | 5 | 4731 | |||
XM_008772428.1 | XP_008770650.1 | 5 | 4487 | |||
XM_008772429.1 | XP_008770651.1 | 7 | 1219 | |||
XM_008772430.1 | XP_008770652.1 | 6 | 1838 | |||
XM_008772431.1 | XP_008770653.1 | 5 | 1731 | |||
XM_008772432.1 | XP_008770654.1 | 6 | 1837 |
