siRNA Details
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100562.1 | NP_001094032.1 | 5, 6 | 478 | ||
XM_006255255.2 | XP_006255317.1 | 5, 6 | 609 | |||
XM_006255257.2 | XP_006255319.1 | 5, 6 | 608 | |||
XM_006255258.2 | XP_006255320.1 | 5, 6 | 610 | |||
XM_008772399.1 | XP_008770621.1 | 5, 6 | 611 | |||
GenBank mRNA | BC099235.1 | - | 16 | |||
CB548211.1 | - | 1, 2 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
