siRNA Details
Gene Symbol: Zcchc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122677.2 | NP_001116149.2 | 13 | 2990 | ||
XM_006249625.2 | XP_006249687.1 | 13 | 2982 | |||
XM_006249626.2 | XP_006249688.1 | 13 | 2982 | |||
XM_006249627.2 | XP_006249689.1 | 12 | 2330 | |||
XM_006249628.2 | XP_006249690.1 | 12 | 2318 |
