siRNA Details
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013125.1 | NP_001013143.1 | 8 | 1444 | ||
GenBank mRNA | BC087103.1 | - | 8 | 1444 | 7192877 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177552, NM_025976, AK028592, BC027221, AK013874.
Gene Ontology Categories:
